Product Details

SNP ID
rs1049474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31879923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCTGGGAGGGAGGGGCTGTCA[A/G]ACCTCCAGGGCCTGGCTGGGATCTC
Phenotype
MIM: 604599 MIM: 606107
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EHMT2 PubMed Links

Gene Details

Gene
EHMT2
Gene Name
euchromatic histone lysine methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289413.1 3888 UTR 3 NP_001276342.1
NM_001318833.1 3888 UTR 3 NP_001305762.1
NM_006709.4 3888 UTR 3 NP_006700.3
NM_025256.6 3888 UTR 3 NP_079532.5
XM_005248824.3 3888 UTR 3 XP_005248881.1
XM_006714974.1 3888 UTR 3 XP_006715037.1
XM_006714975.1 3888 UTR 3 XP_006715038.1
XM_006714976.1 3888 UTR 3 XP_006715039.1
XM_017010202.1 3888 UTR 3 XP_016865691.1
Gene
SLC44A4
Gene Name
solute carrier family 44 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178044.1 3888 Intron NP_001171515.1
NM_001178045.1 3888 Intron NP_001171516.1
NM_025257.2 3888 Intron NP_079533.2

View Full Product Details