Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001197294.1 | 3873 | UTR 3 | NP_001184223.1 | ||
NM_001387.2 | 3873 | UTR 3 | NP_001378.1 | ||
XM_011537574.1 | 3873 | UTR 3 | XP_011535876.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001112724.1 | 3873 | Intron | NP_001106195.1 | ||
NM_001287740.1 | 3873 | Intron | NP_001274669.1 | ||
NM_145001.3 | 3873 | Intron | NP_659438.1 | ||
XM_011537576.1 | 3873 | Intron | XP_011535878.1 | ||
XM_011537577.2 | 3873 | Intron | XP_011535879.1 | ||
XM_011537579.2 | 3873 | Intron | XP_011535881.1 | ||
XM_017009213.1 | 3873 | Intron | XP_016864702.1 | ||
XM_017009214.1 | 3873 | Intron | XP_016864703.1 |