Product Details

SNP ID
rs870856
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:90023055 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAAGGGGACTCTCATCCTGGAATGG[G/C]AGACTCGGTCTGTTGGATCCCTGGG
Phenotype
MIM: 605178 MIM: 605179
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
DBNDD1 PubMed Links

Gene Details

Gene
DBNDD1
Gene Name
dysbindin domain containing 1
There are no transcripts associated with this gene.

Gene
GAS8
Gene Name
growth arrest specific 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286205.1 Intron NP_001273134.1
NM_001286208.1 Intron NP_001273137.1
NM_001286209.1 Intron NP_001273138.1
NM_001481.2 Intron NP_001472.1
XM_005256304.4 Intron XP_005256361.1
XM_005256309.4 Intron XP_005256366.1
XM_006721175.3 Intron XP_006721238.1
XM_011522990.2 Intron XP_011521292.1
XM_011522991.1 Intron XP_011521293.1
XM_011522992.2 Intron XP_011521294.1
XM_017023122.1 Intron XP_016878611.1
XM_017023123.1 Intron XP_016878612.1
XM_017023124.1 Intron XP_016878613.1
XM_017023125.1 Intron XP_016878614.1
Gene
GAS8-AS1
Gene Name
GAS8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details