Product Details

SNP ID
rs1697914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:112712022 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGATTAAACAAACATAGATCAT[A/G]CTGCCAAACCTGTAACAATATCTTT
Phenotype
MIM: 611731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APC PubMed Links

Gene Details

Gene
APC
Gene Name
APC, WNT signaling pathway regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000038.5 Intron NP_000029.2
NM_001127510.2 Intron NP_001120982.1
NM_001127511.2 Intron NP_001120983.2

View Full Product Details