Product Details

SNP ID
rs303448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:30438155 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTTCTAGAAGGCCATTTATTTA[G/T]ATATTAATCTTCTGGGACCCTCAAG
Phenotype
MIM: 191195
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP3K8 PubMed Links

Gene Details

Gene
MAP3K8
Gene Name
mitogen-activated protein kinase kinase kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244134.1 Intron NP_001231063.1
NM_001320961.1 Intron NP_001307890.1
NM_005204.3 Intron NP_005195.2
XM_017015707.1 Intron XP_016871196.1
XM_017015708.1 Intron XP_016871197.1
XM_017015709.1 Intron XP_016871198.1
XM_017015710.1 Intron XP_016871199.1
XM_017015711.1 Intron XP_016871200.1
XM_017015712.1 Intron XP_016871201.1
XM_017015713.1 Intron XP_016871202.1
XM_017015714.1 Intron XP_016871203.1

View Full Product Details