Product Details

SNP ID
rs1063205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:30015989 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGAAGAGGATCTTTTTGTTTCC[C/T]GCACTTATTGCTGCTGTGAAAACTG
Phenotype
MIM: 614398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1462 PubMed Links

Gene Details

Gene
KIAA1462
Gene Name
KIAA1462
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020848.2 6178 UTR 3 NP_065899.1
XM_011519608.1 6178 UTR 3 XP_011517910.1
XM_011519609.2 6178 UTR 3 XP_011517911.1

View Full Product Details