Product Details
- SNP ID
-
rs1493547
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:129380632 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGCCAGAGGCACATAGGCTTTTA[G/T]TAGCAATTACTCATAACTGCTTCTC
- Phenotype
-
MIM: 604823
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BARX2
PubMed Links
Gene Details
- Gene
- BARX2
- Gene Name
- BARX homeobox 2
View Full Product Details