Product Details

SNP ID
rs9822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112063783 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGAAGGAAGGAGGCGATATTCTG[C/G]ATTATAAAAATGAATTGGGAACATT
Phenotype
MIM: 608770
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DLAT PubMed Links

Gene Details

Gene
DLAT
Gene Name
dihydrolipoamide S-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001931.4 3851 UTR 3 NP_001922.2
XM_011542647.2 3851 UTR 3 XP_011540949.1
Gene
PIH1D2
Gene Name
PIH1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082619.1 3851 Intron NP_001076088.1
NM_138789.3 3851 Intron NP_620144.1
XM_011542593.2 3851 Intron XP_011540895.2
XM_017017201.1 3851 Intron XP_016872690.1
XM_017017202.1 3851 Intron XP_016872691.1
XM_017017203.1 3851 Intron XP_016872692.1
XM_017017204.1 3851 Intron XP_016872693.1
XM_017017205.1 3851 Intron XP_016872694.1

View Full Product Details