Product Details

SNP ID
rs797935
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:86922875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGATCTGTGAGTTTACAGTTTT[C/T]ATTACATTTAAACATTTTTGGCTAT
Phenotype
MIM: 604844 MIM: 606254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HS2ST1 PubMed Links

Gene Details

Gene
HS2ST1
Gene Name
heparan sulfate 2-O-sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134492.1 Intron NP_001127964.1
NM_012262.3 Intron NP_036394.1
Gene
SEP15
Gene Name
15 kDa selenoprotein
There are no transcripts associated with this gene.

View Full Product Details