Product Details
- SNP ID
-
rs603301
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:75614009 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTTGTCTTTCTTGACCTTGACAT[A/G]TTTTAAGAAAACAGGGTGGTTATTT
- Phenotype
-
MIM: 605003
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SENP6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78895036] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SENP6
- Gene Name
- SUMO1/sentrin specific peptidase 6
View Full Product Details