Product Details
- SNP ID
-
rs1208606
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:37952139 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCCAACTCATCTTACTTCTCAGC[A/C]TTCCTCTTCTTTGTGTGTGTCTTCT
- Phenotype
-
MIM: 194528
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147942111] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF25
- Gene Name
- zinc finger protein 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
XM_005252384.2 |
1702 |
Missense Mutation |
AAG,AAT |
K453N |
XP_005252441.1 |
XM_005252385.2 |
1702 |
Missense Mutation |
AAG,AAT |
K453N |
XP_005252442.1 |
XM_005252386.2 |
1702 |
Missense Mutation |
AAG,AAT |
K453N |
XP_005252443.1 |
XM_011519352.2 |
1702 |
Missense Mutation |
AAG,AAT |
K457N |
XP_011517654.1 |
XM_011519353.2 |
1702 |
Missense Mutation |
AAG,AAT |
K457N |
XP_011517655.1 |
XM_011519354.2 |
1702 |
Missense Mutation |
AAG,AAT |
K457N |
XP_011517656.1 |
XM_011519355.2 |
1702 |
Missense Mutation |
AAG,AAT |
K457N |
XP_011517657.1 |
XM_011519356.2 |
1702 |
Missense Mutation |
AAG,AAT |
K417N |
XP_011517658.1 |
XM_017015837.1 |
1702 |
Missense Mutation |
AAG,AAT |
K417N |
XP_016871326.1 |
XM_017015838.1 |
1702 |
Missense Mutation |
AAG,AAT |
K417N |
XP_016871327.1 |
View Full Product Details