Product Details

SNP ID
rs1047973
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:184791201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGCAAACCCTCAAACCCGTCT[C/T]TTTATGGTAAAGTGTGAAGGCACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM129A PubMed Links

Gene Details

Gene
FAM129A
Gene Name
family with sequence similarity 129 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052966.3 7557 UTR 3 NP_443198.1
XM_011509140.2 7557 UTR 3 XP_011507442.1
XM_011509141.2 7557 UTR 3 XP_011507443.1

View Full Product Details