Product Details

SNP ID
rs1477467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:155655165 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTATATCCTGGGCACACGTGTACA[C/T]GTATGTTTTACATACACAGTGGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM33 PubMed Links

Gene Details

Gene
RBM33
Gene Name
RNA binding motif protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053043.2 Intron NP_444271.2
XM_005249531.1 Intron XP_005249588.1
XM_005249532.1 Intron XP_005249589.1
XM_005249533.1 Intron XP_005249590.1
XM_005249534.1 Intron XP_005249591.1
XM_005249535.1 Intron XP_005249592.1
XM_011515840.1 Intron XP_011514142.1

View Full Product Details