Product Details

SNP ID
rs1326126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:109381503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTCATTCAGAAATAGATAAATA[C/G]AACACAGTGGAAATGGCAAAAAAAG
Phenotype
MIM: 300041
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GUCY2F PubMed Links

Gene Details

Gene
GUCY2F
Gene Name
guanylate cyclase 2F, retinal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001522.2 Intron NP_001513.2

View Full Product Details