Product Details

SNP ID
rs974570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50191684 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTGACTGACGCAGACGCTTCAA[A/G]ACAAAACCACAATACCTTGCACGAC
Phenotype
MIM: 607917
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SELO PubMed Links

Gene Details

Gene
SELO
Gene Name
selenoprotein O
There are no transcripts associated with this gene.

Gene
TRABD
Gene Name
TraB domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320484.1 Intron NP_001307413.1
NM_001320485.1 Intron NP_001307414.1
NM_001320487.1 Intron NP_001307416.1
NM_001320488.1 Intron NP_001307417.1
NM_025204.3 Intron NP_079480.2
XM_006724422.3 Intron XP_006724485.1
XM_006724423.3 Intron XP_006724486.1
XM_011530712.2 Intron XP_011529014.2
XM_011530713.2 Intron XP_011529015.2
XM_011530715.2 Intron XP_011529017.2
XM_011530716.2 Intron XP_011529018.1
XM_011530717.2 Intron XP_011529019.1
XM_011530718.2 Intron XP_011529020.1
XM_017028943.1 Intron XP_016884432.1
XM_017028944.1 Intron XP_016884433.1

View Full Product Details