Product Details

SNP ID
rs1042979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50275437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTAAGGCACATTTTCAAATCACT[C/G]GAGACTCGACAGTGAACACCCGATG
Phenotype
MIM: 602898 MIM: 604293
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAPK11 PubMed Links

Gene Details

Gene
MAPK11
Gene Name
mitogen-activated protein kinase 11
There are no transcripts associated with this gene.

Gene
PLXNB2
Gene Name
plexin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012401.3 5877 UTR 3 NP_036533.2
XM_005261909.1 5877 UTR 3 XP_005261966.1
XM_005261910.2 5877 UTR 3 XP_005261967.1
XM_005261911.4 5877 UTR 3 XP_005261968.1
XM_006724413.2 5877 UTR 3 XP_006724476.1
XM_011530682.2 5877 UTR 3 XP_011528984.1
XM_017028703.1 5877 UTR 3 XP_016884192.1
XM_017028704.1 5877 UTR 3 XP_016884193.1

View Full Product Details