Product Details

SNP ID
rs1492479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112141699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGAAATAAATAACTTTCACAT[A/C]ATTTATATTTCAATAATGTTTAATA
Phenotype
MIM: 607792 MIM: 612738
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GCSAM PubMed Links

Gene Details

Gene
GCSAM
Gene Name
germinal center associated signaling and motility
There are no transcripts associated with this gene.

Gene
SLC9C1
Gene Name
solute carrier family 9 member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320531.1 Intron NP_001307460.1
NM_183061.2 Intron NP_898884.1
XM_006713603.2 Intron XP_006713666.1
XM_011512718.1 Intron XP_011511020.1
XM_011512719.1 Intron XP_011511021.1
XM_011512720.1 Intron XP_011511022.1
XM_011512721.1 Intron XP_011511023.1
XM_011512724.1 Intron XP_011511026.1
XM_011512725.2 Intron XP_011511027.1
XM_011512726.2 Intron XP_011511028.1
XM_011512727.2 Intron XP_011511029.1
XM_017006246.1 Intron XP_016861735.1
XM_017006247.1 Intron XP_016861736.1
XM_017006248.1 Intron XP_016861737.1
XM_017006249.1 Intron XP_016861738.1
XM_017006250.1 Intron XP_016861739.1

View Full Product Details