Product Details

SNP ID
rs1056464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5044829 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTAAAGAAGAGGAGAAGCCCCTG[C/T]CCCGCAACGAGTTCCAGCGCCAGGT
Phenotype
MIM: 176267
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNA5 PubMed Links

Gene Details

Gene
KCNA5
Gene Name
potassium voltage-gated channel subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002234.3 911 Missense Mutation CCC,TCC P228S NP_002225.2

View Full Product Details