Product Details
- SNP ID
-
rs1399246
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70157397 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTAAAATATCTCAAAGACCCTGA[G/T]GGACAATCTACATTTATTGAGAAAC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRR27
PubMed Links
Gene Details
- Gene
- PRR27
- Gene Name
- proline rich 27
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_214711.3 |
|
Intron |
|
|
NP_999876.2 |
View Full Product Details