Product Details

SNP ID
rs1503860
Assay Type
Functionally Tested
NCBI dbSNP Submissions
45
Location
Chr.1:160543666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTGTTCTCTCAGTCTAACTGGG[A/G]AGGCAGATATAGCATCAGGTAAGTA
Phenotype
MIM: 604513
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CD84 PubMed Links

Gene Details

Gene
CD84
Gene Name
CD84 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184879.1 5346 UTR 3 NP_001171808.1
NM_001184881.1 5346 UTR 3 NP_001171810.1
NM_001184882.1 5346 UTR 3 NP_001171811.1
NM_003874.3 5346 UTR 3 NP_003865.1
XM_011510094.2 5346 UTR 3 XP_011508396.1
XM_011510095.2 5346 UTR 3 XP_011508397.1

View Full Product Details