Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_198468.2 | Intron | NP_940870.2 | |||
XM_006715430.1 | Intron | XP_006715493.1 | |||
XM_006715432.3 | Intron | XP_006715495.1 | |||
XM_011535670.2 | Intron | XP_011533972.1 | |||
XM_011535671.2 | Intron | XP_011533973.1 | |||
XM_011535672.2 | Intron | XP_011533974.1 | |||
XM_011535673.2 | Intron | XP_011533975.1 | |||
XM_011535674.2 | Intron | XP_011533976.1 | |||
XM_011535675.2 | Intron | XP_011533977.1 | |||
XM_011535676.2 | Intron | XP_011533978.1 | |||
XM_011535677.2 | Intron | XP_011533979.1 | |||
XM_011535678.2 | Intron | XP_011533980.1 | |||
XM_011535679.2 | Intron | XP_011533981.1 | |||
XM_011535680.2 | Intron | XP_011533982.1 | |||
XM_017010656.1 | Intron | XP_016866145.1 | |||
XM_017010657.1 | Intron | XP_016866146.1 |