Product Details
- SNP ID
-
rs1629534
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1809481 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGCGACTCGTGCTGGCCGAGCCC[A/C]GCCCTCAGAGGACACGGAGGGCGGG
- Phenotype
-
MIM: 138760
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HAGH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs118097552] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAGH
- Gene Name
- hydroxyacylglutathione hydrolase
View Full Product Details