Product Details

SNP ID
rs1042138
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:93426338 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAATGATGGCTCAGTGATCACGAT[A/G]CTGTGTTTGCTTCACATTCAAGCAG
Phenotype
MIM: 114131
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALCR PubMed Links

Gene Details

Gene
CALCR
Gene Name
calcitonin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164737.1 1726 UTR 3 NP_001158209.1
NM_001164738.1 1726 UTR 3 NP_001158210.1
NM_001742.3 1726 UTR 3 NP_001733.1

View Full Product Details