Product Details

SNP ID
rs974560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38962555 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCCTCTGTGCGGGGGGAGCTCTGG[C/T]TCAGGCTCCCCAGTGGTCCCGAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX35 PubMed Links

Gene Details

Gene
DHX35
Gene Name
DEAH-box helicase 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190809.1 Intron NP_001177738.1
NM_021931.3 Intron NP_068750.2
XM_006723848.3 Intron XP_006723911.1
XM_011528972.1 Intron XP_011527274.1
XM_011528974.1 Intron XP_011527276.1
XM_017028004.1 Intron XP_016883493.1
Gene
FAM83D
Gene Name
family with sequence similarity 83 member D
There are no transcripts associated with this gene.

View Full Product Details