Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001136021.2 | 3910 | UTR 3 | NP_001129493.1 | ||
NM_001258292.1 | 3910 | UTR 3 | NP_001245221.1 | ||
NM_001258294.1 | 3910 | UTR 3 | NP_001245223.1 | ||
NM_001258295.1 | 3910 | UTR 3 | NP_001245224.1 | ||
NM_001258296.1 | 3910 | UTR 3 | NP_001245225.1 | ||
NM_001258297.1 | 3910 | UTR 3 | NP_001245226.1 | ||
NM_012340.4 | 3910 | UTR 3 | NP_036472.2 | ||
NM_173091.3 | 3910 | UTR 3 | NP_775114.1 | ||
XM_011528824.2 | 3910 | Intron | XP_011527126.1 | ||
XM_011528825.2 | 3910 | Intron | XP_011527127.1 | ||
XM_011528826.2 | 3910 | Intron | XP_011527128.1 | ||
XM_017027850.1 | 3910 | Intron | XP_016883339.1 | ||
XM_017027851.1 | 3910 | UTR 3 | XP_016883340.1 |