Product Details

SNP ID
rs1446150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74177698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATGTCAAATGTCAAGTCGTTTA[A/G]GTTTGTTATCCTTCTTCACTCCAAG
Phenotype
MIM: 603431
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4H PubMed Links

Gene Details

Gene
EIF4H
Gene Name
eukaryotic translation initiation factor 4H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022170.1 Intron NP_071496.1
NM_031992.1 Intron NP_114381.1
XM_011516556.1 Intron XP_011514858.1
XM_011516557.1 Intron XP_011514859.1

View Full Product Details