Product Details

SNP ID
rs1272909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59475647 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAATAATGTCATTATTATGAACA[C/T]AGTTTTGACCCTCTGAACCCATGGA
Phenotype
MIM: 611176 MIM: 614811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JKAMP PubMed Links

Gene Details

Gene
JKAMP
Gene Name
JNK1/MAPK8-associated membrane protein
There are no transcripts associated with this gene.

Gene
L3HYPDH
Gene Name
trans-L-3-hydroxyproline dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144581.1 Intron NP_653182.1
XM_005267298.3 Intron XP_005267355.1
XM_011536386.2 Intron XP_011534688.1
XM_011536387.1 Intron XP_011534689.1
XM_011536388.1 Intron XP_011534690.1

View Full Product Details