Product Details

SNP ID
rs888818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80961718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACGCTTCTGTACTTTTAAATGA[A/G]TGCTTTGAAAATGAATGCGACCTTT
Phenotype
MIM: 606614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASGRF2 PubMed Links

Gene Details

Gene
RASGRF2
Gene Name
Ras protein specific guanine nucleotide releasing factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006909.2 Intron NP_008840.1
XM_005248565.1 Intron XP_005248622.1
XM_017009682.1 Intron XP_016865171.1
XM_017009683.1 Intron XP_016865172.1
Gene
RASGRF2-AS1
Gene Name
RASGRF2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details