Product Details

SNP ID
rs974831
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:102285836 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGAGAAGACACCCAGGGGTCCCCCC[A/C]TGCTATCTCCAGGAGCAAGACTACT
Phenotype
MIM: 116896 MIM: 605300
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CUX1 PubMed Links

Gene Details

Gene
CUX1
Gene Name
cut like homeobox 1
There are no transcripts associated with this gene.

Gene
MIR4285
Gene Name
microRNA 4285
There are no transcripts associated with this gene.

Gene
SH2B2
Gene Name
SH2B adaptor protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020979.4 Intron NP_066189.3
XM_005276975.4 Intron XP_005277032.1
XM_005276976.3 Intron XP_005277033.1
XM_005276977.4 Intron XP_005277034.1
XM_005276979.3 Intron XP_005277036.1
XM_011515740.2 Intron XP_011514042.1
XM_017011680.1 Intron XP_016867169.1
XM_017011681.1 Intron XP_016867170.1

View Full Product Details