Product Details

SNP ID
rs1803500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124619418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGCTGCAGGGGCGGGCAGAC[C/T]CTGGGCCCCGGGTCTACGCTTTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM53B PubMed Links

Gene Details

Gene
FAM53B
Gene Name
family with sequence similarity 53 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014661.3 5616 UTR 3 NP_055476.3
Gene
LHPP
Gene Name
phospholysine phosphohistidine inorganic pyrophosphate phosphatase
There are no transcripts associated with this gene.

View Full Product Details