Product Details

SNP ID
rs1439795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:131111075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTCCAGACAACTTTTTAATTA[C/T]GATTATTCCCTTCCATTGCACTCCT
Phenotype
MIM: 605623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC-PINT PubMed Links

Gene Details

Gene
LINC-PINT
Gene Name
long intergenic non-protein coding RNA, p53 induced transcript
There are no transcripts associated with this gene.

Gene
MKLN1
Gene Name
muskelin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145354.1 Intron NP_001138826.1
NM_001321316.1 Intron NP_001308245.1
NM_013255.4 Intron NP_037387.2
XM_006715993.2 Intron XP_006716056.1
XM_011516224.2 Intron XP_011514526.1

View Full Product Details