Product Details
- SNP ID
-
rs8523
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:10980820 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGATAATGCTTTTTATCTACAAAGA[G/A]AAATGGCTTCTGCAGCCTCCCTGTC
- Phenotype
-
MIM: 611814
MIM: 616799
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ELOVL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41507245] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ELOVL2
- Gene Name
- ELOVL fatty acid elongase 2
- Gene
- SYCP2L
- Gene Name
- synaptonemal complex protein 2 like
There are no transcripts associated with this gene.
View Full Product Details