Product Details

SNP ID
rs409056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:182717292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTTTTGTTTCTACTTACACCA[A/C]ACATGAACTTTGCAATAAAATTACA
Phenotype
MIM: 607987
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC10 PubMed Links

Gene Details

Gene
DNAJC10
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271581.1 Intron NP_001258510.1
NM_018981.2 Intron NP_061854.1

View Full Product Details