Product Details
- SNP ID
-
rs8217
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:129835456 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGAGACAGTGAAGTGATGTGGGCA[C/T]TGACTACTTAACCTAAAACACAAAC
- Phenotype
-
MIM: 607407
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EBF3
PubMed Links
Gene Details
- Gene
- EBF3
- Gene Name
- early B-cell factor 3
- Gene
- MIR4297
- Gene Name
- microRNA 4297
There are no transcripts associated with this gene.
View Full Product Details