Product Details
- SNP ID
-
rs995202
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:127551661 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATCATGGGTAAATCTGTGCTTTGC[A/C]TTATTGGTGAAATTTGTATTAGCAT
- Phenotype
-
MIM: 609513
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NPS
PubMed Links
Gene Details
- Gene
- NPS
- Gene Name
- neuropeptide S
View Full Product Details