Product Details
- SNP ID
-
rs14332
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:17179611 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGACAAAAGGGGAAAGAAGGAAT[C/G]TTCCAGGCAGAGAGAAAGAGAAAAG
- Phenotype
-
MIM: 607575
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CECR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76206943] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CECR1
- Gene Name
- cat eye syndrome chromosome region, candidate 1
View Full Product Details