Product Details

SNP ID
rs1533764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:76189955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCAGGAAACCTGTGGGGGCTG[C/T]GAAGGAAGCCAGGAGGGTAAGTGGT
Phenotype
MIM: 603699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WNT11 PubMed Links

Gene Details

Gene
WNT11
Gene Name
Wnt family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004626.2 Intron NP_004617.2
XM_005274231.1 Intron XP_005274288.1
XM_011545238.1 Intron XP_011543540.1
XM_011545239.2 Intron XP_011543541.1
XM_011545240.2 Intron XP_011543542.1
XM_011545241.2 Intron XP_011543543.1
XM_017018241.1 Intron XP_016873730.1
XM_017018242.1 Intron XP_016873731.1
XM_017018243.1 Intron XP_016873732.1

View Full Product Details