Product Details

SNP ID
rs17067906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138807446 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACACATTTTCTATCCTCCACATTA[C/G]TGCAGGTAACAGTGTTACTAACTTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ECT2L PubMed Links

Gene Details

Gene
ECT2L
Gene Name
epithelial cell transforming 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077706.2 Intron NP_001071174.1
NM_001195037.2 Intron NP_001181966.1
XM_006715472.3 Intron XP_006715535.1
XM_011535795.2 Intron XP_011534097.1
XM_011535797.2 Intron XP_011534099.1
XM_017010828.1 Intron XP_016866317.1
XM_017010829.1 Intron XP_016866318.1
XM_017010830.1 Intron XP_016866319.1

View Full Product Details