Product Details

SNP ID
rs11752519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158849599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGGCTTTTAGCTGGCATTAGG[A/G]GTTTGAGACCTTGGAATGTTGTCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSTCP1 PubMed Links

Gene Details

Gene
OSTCP1
Gene Name
oligosaccharyltransferase complex subunit pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details