Product Details

SNP ID
rs3732793
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:113723351 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTTAAAAAGACCCAAATTAACTTA[A/C]TAGGTCAGACACATTAACAAATAAT
Phenotype
MIM: 610834
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAA50 PubMed Links

Gene Details

Gene
NAA50
Gene Name
N(alpha)-acetyltransferase 50, NatE catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308445.1 Intron NP_001295374.1
NM_025146.3 Intron NP_079422.1

View Full Product Details