Product Details
- SNP ID
-
rs10414406
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:45082596 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAGGCACTCAGATGATGTGCTAAC[A/G]ATGCTAGATAATGTGGATGAGTGAA
- Phenotype
-
MIM: 607419
MIM: 613226
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GEMIN7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10416359] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GEMIN7
- Gene Name
- gem nuclear organelle associated protein 7
- Gene
- LOC105372419
- Gene Name
- uncharacterized LOC105372419
There are no transcripts associated with this gene.
- Gene
- ZNF296
- Gene Name
- zinc finger protein 296
There are no transcripts associated with this gene.
View Full Product Details