Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002000.3 | 56 | UTR 5 | NP_001991.1 | ||
NM_133269.3 | 56 | UTR 5 | NP_579803.1 | ||
NM_133271.3 | 56 | UTR 5 | NP_579805.1 | ||
NM_133272.3 | 56 | UTR 5 | NP_579806.1 | ||
NM_133273.3 | 56 | UTR 5 | NP_579807.1 | ||
NM_133274.3 | 56 | UTR 5 | NP_579808.1 | ||
NM_133277.3 | 56 | UTR 5 | NP_579811.1 | ||
NM_133278.3 | 56 | UTR 5 | NP_579812.1 | ||
XM_011526625.2 | 56 | Intron | XP_011524927.1 | ||
XM_017026473.1 | 56 | Intron | XP_016881962.1 | ||
XM_017026474.1 | 56 | Intron | XP_016881963.1 |