Product Details
- SNP ID
-
rs11794604
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:112692097 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGTCTGATAAGAAAAGGACTCTGG[C/T]CTGCCAGATTTAACCCGGGAAGGTT
- Phenotype
-
MIM: 613273
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
INIP
PubMed Links
Gene Details
- Gene
- INIP
- Gene Name
- INTS3 and NABP interacting protein
There are no transcripts associated with this gene.
View Full Product Details