Product Details

SNP ID
rs11672506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:29534826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCAGTACATCTCCCCAAATAGT[C/G]CCCCATTAAGCGATCGTTCCTGGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC284395 PubMed Links

Gene Details

Gene
LOC284395
Gene Name
uncharacterized LOC284395
There are no transcripts associated with this gene.

Gene
VSTM2B
Gene Name
V-set and transmembrane domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146339.1 Intron NP_001139811.1
XM_005258856.3 Intron XP_005258913.1
XM_011526902.2 Intron XP_011525204.1
XM_011526903.2 Intron XP_011525205.1
XM_011526904.2 Intron XP_011525206.1
XM_011526905.2 Intron XP_011525207.1

View Full Product Details