Product Details
- SNP ID
-
rs1893032
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:119218173 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACAGTCCCACTTAACATAAATGTA[A/G]TAGTTGCTGGGGAGTTAAAGATGAC
- Phenotype
-
MIM: 165360
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBL
PubMed Links
Gene Details
- Gene
- CBL
- Gene Name
- Cbl proto-oncogene
View Full Product Details