Product Details

SNP ID
rs1533134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139111511 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGCTGGGATTTTTCAGGCATCA[C/T]AGCCAGAGACCTTCTGGAAAAGGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPESC1 PubMed Links

Gene Details

Gene
BPESC1
Gene Name
blepharophimosis, epicanthus inversus and ptosis, candidate 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details