Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130864.1 | 1953 | Intron | NP_001124336.1 | ||
NM_001267035.1 | 1953 | Intron | NP_001253964.1 | ||
NM_052927.2 | 1953 | Intron | NP_443159.1 | ||
XM_005265811.3 | 1953 | Intron | XP_005265868.1 | ||
XM_005265812.2 | 1953 | Intron | XP_005265869.1 | ||
XM_006714817.3 | 1953 | Intron | XP_006714880.1 | ||
XM_011534424.2 | 1953 | Intron | XP_011532726.1 | ||
XM_017009009.1 | 1953 | Intron | XP_016864498.1 | ||
XM_017009010.1 | 1953 | Intron | XP_016864499.1 | ||
XM_017009011.1 | 1953 | Intron | XP_016864500.1 | ||
XM_017009012.1 | 1953 | UTR 3 | XP_016864501.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282500.1 | 1953 | Intron | NP_001269429.1 | ||
NM_003314.2 | 1953 | Intron | NP_003305.1 |