Product Details

SNP ID
rs768995
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118903110 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAGATGTTTTATATTTACAACA[G/T]ATTTTAAAACAGAAACATGCCTAGA
Phenotype
MIM: 609189 MIM: 610098
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ASF1A PubMed Links
Additional Information
For this assay, SNP(s) [rs147487888] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ASF1A
Gene Name
anti-silencing function 1A histone chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014034.2 Intron NP_054753.1
Gene
MCM9
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017696.2 Intron NP_060166.2
NM_153255.4 Intron NP_694987.1

View Full Product Details