Product Details
- SNP ID
-
rs776826
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70000504 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGAAGAAACACCACATAAGCTGAA[A/T]TTAGTAAATGTCTCCTTCCCATAAT
- Phenotype
-
MIM: 184470
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
STATH
PubMed Links
Gene Details
- Gene
- STATH
- Gene Name
- statherin
View Full Product Details