Product Details
- SNP ID
-
rs776825
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:69998122 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCACCCAACACCTCTTTACTCATCC[C/T]GCATAATCCAACTCAAGCTTTACTT
- Phenotype
-
MIM: 184470
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STATH
PubMed Links
Gene Details
- Gene
- STATH
- Gene Name
- statherin
View Full Product Details